Methyl 8-azabicyclo[3.2.1]octane-3-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A260181-100MG A260181-1G A260181-250MG A260181-5G
76815-996EA 49 USD
76815-996 76815-998 76816-000 77883-716
Methyl 8-azabicyclo[3.2.1]octane-3-carboxylate hydrochloride 95%
Methyl 8-azabicyclo[3.2.1]octane-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆ClNO₂
MDL Number: MFCD19707653
CAS Number: 179022-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR