H-β-HomoAla-OH·HCl 96%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Aminobutanoic acid hydrochloride

A259316-1G A259316-250MG
76815-970EA 47.75 USD
76815-970 76815-972
H-β-HomoAla-OH·HCl 96%
H-β-HomoAla-OH·HCl

Precaución: for Research Use Only

Formula: C₄H₉NO₂·HCl
MW: 139.6 g/mol
MDL Number: MFCD01862873
CAS Number: 58610-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR