o-Anisic acid 98%

Supplier: Ambeed
Warning

A259232-100G A259232-25G A259232-500G A259232-1000G
76815-964EA 8 USD
76815-964 76815-966 76815-968 77672-310
o-Anisic acid 98%
o-Anisic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00002431
CAS Number: 579-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR