Methyl 2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A258354-100MG A258354-250MG
76815-956EA 108.22 USD
76815-956 76815-958
Methyl 2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate hydrochloride 95%
Methyl 2-(2-chlorophenyl)-2-(4,5-dihydrothieno[2,3-c]pyridin-6(7H)-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₇Cl₂NO₂S
MW: 358.28 g/mol
MDL Number: MFCD22375692
CAS Number: 144750-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR