3-Chloropropylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloropropan-1-amine hydrochloride

A257632-1000G A257632-100G A257632-10G A257632-25G A257632-500G A257632-5G
76815-924EA 89.12 USD
76815-924 76815-926 76815-928 76815-930 76815-932 76815-934
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₈ClN
MW: 130.01 g/mol
MDL Number: MFCD00012913
CAS Number: 6276-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR