Methyl 3-amino-4-bromothiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A257569-10G A257569-1G A257569-250MG A257569-25G A257569-100MG
76815-916EA 306.61 USD
76815-916 76815-918 76815-920 76815-922 77672-298
Methyl 3-amino-4-bromothiophene-2-carboxylate 95%
Methyl 3-amino-4-bromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.09 g/mol
MDL Number: MFCD09038329
CAS Number: 161833-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR