Pyrazolo[1,5-a]pyrimidin-5(1H)-one 95%

Supplier: Ambeed
Warning

A257427-1G A257427-250MG A257427-5G
76815-906EA 37.14 USD
76815-906 76815-908 76815-910
Pyrazolo[1,5-a]pyrimidin-5(1H)-one 95%
Pyrazolo[1,5-a]pyrimidin-5(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅N₃O
MW: 135.12 g/mol
MDL Number: MFCD27989151
CAS Number: 1027534-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR