Ethyl-1-phenyl-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-phenyl-1H-pyrazole-4-carboxylate

A257373-10G A257373-1G A257373-5G
76815-900EA 233.4 USD
76815-900 76815-902 76815-904
Ethyl-1-phenyl-1H-pyrazole-4-carboxylate 97%
Ethyl-1-phenyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.2400 g/mol
MDL Number: MFCD00973358
CAS Number: 885-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR