(Z)-5-((Z)-3-Chloro-4-((R)-2,3-dihydroxypropoxy)benzylidene)-2-(propylimino)-3-(o-tolyl)thiazolidin-4-one 98%

Supplier: Ambeed
Warning

A257238-10MG A257238-1MG A257238-5MG A257238-1G A257238-250MG A257238-100MG
76815-892EA 43 USD
76815-892 76815-894 76815-896 76815-898 77672-292 77672-294 77883-700
(Z)-5-((Z)-3-Chloro-4-((R)-2,3-dihydroxypropoxy)benzylidene)-2-(propylimino)-3-(o-tolyl)thiazolidin-4-one 98%
(Z)-5-((Z)-3-Chloro-4-((R)-2,3-dihydroxypropoxy)benzylidene)-2-(propylimino)-3-(o-tolyl)thiazolidin-4-one

Precaución: for Research Use Only

Formula: C₂₃H₂₅ClN₂O₄S
MDL Number: MFCD18207776
CAS Number: 854107-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR