N-Cyclopropyl 4-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Cyclopropylsulphamoyl)benzeneboronic acid, (4-(N-Cyclopropylsulphamoyl)phenyl)boronic acid

A257100-100MG A257100-1G A257100-250MG A257100-5G
76815-878EA 36.08 USD
76815-878 76815-880 76815-882 76815-884
N-Cyclopropyl 4-boronobenzenesulfonamide 98%
N-Cyclopropyl 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₄S
MW: 241.07 g/mol
MDL Number: MFCD07363755
CAS Number: 871329-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR