2,6-Dibromophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2,6-Dibromophenyl)boronic acid

A256202-1G A256202-250MG
76815-860EA 109.28 USD
76815-860 76815-862
2,6-Dibromophenylboronic acid 95%
2,6-Dibromophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBr₂O₂
MW: 279.72 g/mol
MDL Number: MFCD11656174
CAS Number: 851756-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR