Berberine chloride 97%

Supplier: Ambeed
Warning

A256069-100MG A256069-10G A256069-25G A256069-500G A256069-5G A256069-100G
76815-848EA 6 USD
76815-848 76815-850 76815-852 76815-854 76815-856 77612-458
Berberine chloride 97%
Berberine chloride

Precaución: for Research Use Only

Formula: C₂₀H₁₈ClNO₄
MDL Number: MFCD00011939
CAS Number: 633-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR