2-Chloro-3-iodoisonicotinonitrile 95%

Supplier: Ambeed
Warning

A255993-100MG A255993-250MG
76815-846EA 42 USD
76815-846 77612-456
2-Chloro-3-iodoisonicotinonitrile 95%
2-Chloro-3-iodoisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂ClIN₂
MDL Number: MFCD09056805
CAS Number: 908279-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR