Di-p-toluoyl-L-tartaric acid monohydrate 99%
Supplier: Ambeed
|
Warning
|
A255766-1000G
A255766-100G
A255766-25G
76815-834EA
72.15
USD
76815-834
76815-836
76815-838
Di-p-toluoyl-L-tartaric acid monohydrate 99%
Di-p-toluoyl-L-tartaric acid monohydrate
Precaución: for Research Use Only
Formula:
C₂₀H₂₀O₉ MW: 404.36 g/mol |
MDL Number:
MFCD00151034 CAS Number: 71607-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...