(R)-2-(4-Hydroxy-6-methylnicotinamido)-2-(4-hydroxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A255435-100G A255435-10G A255435-1G A255435-250MG A255435-25G A255435-5G
76815-814EA 78.51 USD
76815-814 76815-816 76815-818 76815-820 76815-822 76815-824
(R)-2-(4-Hydroxy-6-methylnicotinamido)-2-(4-hydroxyphenyl)acetic acid 95%
(R)-2-(4-Hydroxy-6-methylnicotinamido)-2-(4-hydroxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₅
MW: 302.28 g/mol
MDL Number: MFCD09033158
CAS Number: 70785-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR