4-Amino-3,5-dichloro-2-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-2-methylpyridin-4-amine

A255164-100MG A255164-1G A255164-250MG
76815-800EA 50.93 USD
76815-800 76815-802 76815-804
4-Amino-3,5-dichloro-2-methylpyridine 97%
4-Amino-3,5-dichloro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.031 g/mol
MDL Number: MFCD00160307
CAS Number: 195045-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR