(3-Formyl-5-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A254605-100MG A254605-1G A254605-250MG A254605-5G
76815-780EA 9 USD
76815-780 76815-782 76815-784 76815-786
(3-Formyl-5-methylphenyl)boronic acid 97%
(3-Formyl-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MDL Number: MFCD08276762
CAS Number: 870777-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR