4-tert-Butyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(tert-Butyl)-1H-pyrazole
A254319-100MG
A254319-250MG
A254319-5G
76815-772EA
22.28
USD
76815-772
76815-774
76815-776
4-tert-Butyl-1H-pyrazole 97%
4-tert-Butyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₇H₁₂N₂ MW: 124.184 g/mol |
MDL Number:
MFCD11847492 CAS Number: 105285-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...