2-Methylpyrimidine-4,6(1H,5H)-dione 97%

Supplier: Ambeed
Warning

A253557-25G A253557-5G
76815-734EA 11.67 USD
76815-734 76815-736
2-Methylpyrimidine-4,6(1H,5H)-dione 97%
2-Methylpyrimidine-4,6(1H,5H)-dione

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD07189479
CAS Number: 40497-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR