(2-Fluoro-4-nitrophenyl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A253340-100MG A253340-250MG
76815-726EA 45.62 USD
76815-726 76815-728
(2-Fluoro-4-nitrophenyl)methanamine hydrochloride 95%
(2-Fluoro-4-nitrophenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClFN₂O₂
MW: 206.60 g/mol
MDL Number: MFCD13194374
CAS Number: 937783-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR