(S)-1-Boc-2-isopropylpiperazine 98%
Supplier: Ambeed
|
Warning
|
A252963-100MG
A252963-1G
A252963-250MG
A252963-5G
76815-714EA
14
USD
76815-714
76815-716
76815-718
77612-436
(S)-1-Boc-2-isopropylpiperazine 98%
(S)-1-Boc-2-isopropylpiperazine
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₂O₂ |
MDL Number:
MFCD03787918 CAS Number: 674792-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...