3,4-Difluoro-5-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-m-tolualdehyde, 3,4-Difluoro-5-tolylaldehyde, 3,4-Difluoro-5-tolualdehyde, 4,5-Difluoro-m-tolylaldehyde

A252869-100MG A252869-1G A252869-250MG
76815-708EA 82.76 USD
76815-708 76815-710 76815-712
3,4-Difluoro-5-methylbenzaldehyde 97%
3,4-Difluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD09832333
CAS Number: 1017778-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR