2,3-Difluoro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A252730-100MG A252730-10G A252730-1G A252730-250MG A252730-5G
76815-698EA 37.14 USD
76815-698 76815-700 76815-702 76815-704 76815-706
2,3-Difluoro-5-nitropyridine 98%
2,3-Difluoro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂F₂N₂O₂
MW: 160.08 g/mol
MDL Number: MFCD09750303
CAS Number: 954219-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR