2,3-Dichloro-5,6-dimethylpyrazine 97%

Supplier: Ambeed
Warning

A252324-100MG A252324-1G A252324-250MG
76815-688EA 20 USD
76815-688 77550-602 77568-446
2,3-Dichloro-5,6-dimethylpyrazine 97%
2,3-Dichloro-5,6-dimethylpyrazine

Precaución: For research use only.

Formula: C₆H₆Cl₂N₂
MDL Number: MFCD18449257
CAS Number: 32493-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR