2-(Methylthio)benzothiazole 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Methylthio)benzo[d]thiazole
A251856-10G
A251856-25G
A251856-5G
A251856-100G
A251856-500G
76815-672EA
11.67
USD
76815-672
76815-674
76815-676
77612-432
77672-254
2-(Methylthio)benzothiazole 96%
2-(Methylthio)benzothiazole
Precaución: for Research Use Only
Formula:
C₈H₇NS₂ MW: 181.2779 g/mol |
MDL Number:
MFCD00005784 CAS Number: 615-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...