2-Chloro-5-phenyloxazole 97%

Supplier: Ambeed
Warning

A251515-1G A251515-5G
76815-664EA 274 USD
76815-664 76815-666
2-Chloro-5-phenyloxazole 97%
2-Chloro-5-phenyloxazole

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.603 g/mol
MDL Number: MFCD08688603
CAS Number: 62124-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR