8-(tert-Butoxycarbonyl)-8-azabicyclo[3.2.1]octane-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A251338-1G A251338-250MG
76815-658EA 219.61 USD
76815-658 76815-660
8-(tert-Butoxycarbonyl)-8-azabicyclo[3.2.1]octane-3-carboxylic acid 97%
8-(tert-Butoxycarbonyl)-8-azabicyclo[3.2.1]octane-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MW: 255.31 g/mol
MDL Number: MFCD12068415
CAS Number: 1159826-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR