2,3-Dihydroxyterephthalaldehyde 95%

Supplier: Ambeed
Warning

A250757-100MG A250757-1G A250757-250MG
76815-648EA 44 USD
76815-648 77883-677 77883-678
2,3-Dihydroxyterephthalaldehyde 95%
2,3-Dihydroxyterephthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₆O₄
MDL Number: MFCD24394548
CAS Number: 148063-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR