3'-Amino-4'-methoxyacetanilide 97%

Supplier: Ambeed
Warning

A250347-10G A250347-5G A250347-100G A250347-25G A250347-500G
76815-638EA 6 USD
76815-638 76815-640 77672-230 77883-673 77883-674
3'-Amino-4'-methoxyacetanilide 97%
3'-Amino-4'-methoxyacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00008676
CAS Number: 6375-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR