Isoamyl 4-methoxycinnamate 96% (mixture of isomers)

Supplier: Ambeed
Warning

A249998-100G A249998-5G A249998-25G
76815-628EA 156 USD
76815-628 76815-630 77883-672
Isoamyl 4-methoxycinnamate 96% (mixture of isomers)
Isoamyl 4-methoxycinnamate

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
MDL Number: MFCD00583856
CAS Number: 71617-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR