Methyl-2-amino-5-iodonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-iodonicotinate

A248967-100MG A248967-1G A248967-250MG
76815-606EA 12.74 USD
76815-606 76815-608 76815-610
Methyl-2-amino-5-iodonicotinate 98%
Methyl-2-amino-5-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₇IN₂O₂
MW: 278.05 g/mol
MDL Number: MFCD20266452
CAS Number: 1227048-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR