Methyl-4-fluoro-3-methoxybenzoate 98%

Supplier: Ambeed
Warning

A248048-250MG A248048-5G A248048-1G
76815-592EA 13 USD
76815-592 76815-594 77672-216
Methyl-4-fluoro-3-methoxybenzoate 98%
Methyl-4-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MDL Number: MFCD06203732
CAS Number: 74385-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR