6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one 95%

Supplier: Ambeed
Warning

A247800-1G A247800-250MG A247800-100MG A247800-50MG
76815-584EA 458 USD
76815-584 76815-586 77672-214 77883-668
6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one 95%
6-Bromo-8-fluoro-3,4-dihydroisoquinolin-1(2h)-one

Precaución: for Research Use Only

Formula: C₉H₇BrFNO
MDL Number: MFCD22381969
CAS Number: 1242157-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR