Ethyl-6-chloropicolinate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate

A247674-100G A247674-1G A247674-25G A247674-5G
76815-574EA 397.84 USD
76815-574 76815-576 76815-578 76815-580
Ethyl-6-chloropicolinate 95%
Ethyl 6-chloropicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD09878438
CAS Number: 21190-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR