4-Chlorothieno[2,3-b]pyridine 96%

Supplier: Ambeed
Warning

A247611-1G A247611-250MG
76815-570EA 269.47 USD
76815-570 76815-572
4-Chlorothieno[2,3-b]pyridine 96%
4-Chlorothieno[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClNS
MW: 169.631 g/mol
MDL Number: MFCD11858513
CAS Number: 62226-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR