8-Methyl-4-quinazolone 97%

Supplier: Ambeed
Warning

Synonyms: 8-Methylquinazolin-4(3H)-one

A247355-5G A247355-1G
76815-568EA 135.8 USD
76815-568 77672-210
8-Methyl-4-quinazolone 97%
8-Methyl-4-quinazolone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD01685945
CAS Number: 19181-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR