4-Bromo-2-methylbutan-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methyl-2-butanol

A247350-250MG A247350-1G
76815-566EA 46.68 USD
76815-566 77568-404
4-Bromo-2-methylbutan-2-ol 98%
4-Bromo-2-methylbutan-2-ol

Precaución: For research use only.

Formula: C₅H₁₁BrO
MW: 167.044 g/mol
MDL Number: MFCD20483824
CAS Number: 35979-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR