tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate 98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl (1R,5S,6s)-rel-3-azabicyclo[3.1.0]hexan-6-ylcarbamate

A247344-1G A247344-5G
76815-556EA 307.67 USD
76815-556 76815-558
tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate 98%
tert-Butyl-N-[(1S,5R)-3-azabicyclo[3.1.0]hexan-6-yl]carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.27 g/mol
MDL Number: MFCD12198677
CAS Number: 134575-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR