3-Chloroazetidine hydrochloride 98%

Supplier: Ambeed
Danger

A247103-5G A247103-1G
76815-548EA 157 USD
76815-548 77672-204
3-Chloroazetidine hydrochloride 98%
3-Chloroazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇Cl₂N
MW: 128 g/mol
MDL Number: MFCD09834116
CAS Number: 313468-63-2
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR