(S)-2-(((Benzyloxy)carbonyl)amino)-4-((tert-butoxycarbonyl)amino)butanoic acid 95%

Supplier: Ambeed
Warning

A246850-100G A246850-10G A246850-1G A246850-25G A246850-5G
76815-532EA 807 USD
76815-532 76815-534 76815-536 76815-538 76815-540
(S)-2-(((Benzyloxy)carbonyl)amino)-4-((tert-butoxycarbonyl)amino)butanoic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)-4-((tert-butoxycarbonyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₆
MDL Number: MFCD01632076
CAS Number: 49855-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR