2-(2-Ethoxyphenoxy)ethylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Aminoethoxy)-2-ethoxybenzene, 2-(2-Ethoxyphenoxy)ethanamine

A246681-100MG A246681-1G A246681-250MG A246681-5G
76815-524EA 51 USD
76815-524 76815-526 76815-528 76815-530
2-(2-Ethoxyphenoxy)ethylamine 95%
2-(2-Ethoxyphenoxy)ethylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD06657756
CAS Number: 6781-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR