2-(Difluoromethoxy)naphthalene 98%

Supplier: Ambeed
Warning

A246641-1G A246641-250MG
76815-520EA 87 USD
76815-520 76815-522
2-(Difluoromethoxy)naphthalene 98%
2-(Difluoromethoxy)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₈F₂O
MDL Number: MFCD05856249
CAS Number: 712-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR