2,5-Diiodothiophene 98%

Supplier: Ambeed
Warning

A246509-1G A246509-25G A246509-5G
76815-514EA 11 USD
76815-514 76815-516 76815-518
2,5-Diiodothiophene 98%
2,5-Diiodothiophene

Precaución: for Research Use Only

Formula: C₄H₂I₂S
MW: 335.93 g/mol
Melting Pt: 45
MDL Number: MFCD00014525
CAS Number: 625-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR