Methyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A244333-100MG A244333-1G A244333-250MG
76815-492EA 81.69 USD
76815-492 76815-494 76815-496
Methyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate 97%
Methyl 5-amino-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD10697500
CAS Number: 92406-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR