6-Bromo-2-methylpyridin-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-hydoxy-2-methylpyridine

A243654-100MG A243654-250MG A243654-5G A243654-1G
76815-478EA 21.22 USD
76815-478 76815-480 77593-552 77612-356
6-Bromo-2-methylpyridin-3-ol 97%
6-Bromo-2-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.022 g/mol
MDL Number: MFCD11044277
CAS Number: 118399-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR