6-Chloro-1,8-naphthyridin-2(1H)-one 96%

Supplier: Ambeed
Warning

A243649-100MG A243649-1G A243649-250MG A243649-5G
76815-472EA 11 USD
76815-472 76815-474 76815-476 77883-643
6-Chloro-1,8-naphthyridin-2(1H)-one 96%
6-Chloro-1,8-naphthyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD18260370
CAS Number: 127446-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR