Pentafluorobenzoic acid 98%

Supplier: Ambeed
Warning

A242355-1000G A242355-100G A242355-10G A242355-500G A242355-25G
76815-454EA 135 USD
76815-454 76815-456 76815-458 76815-460 77090-452
Pentafluorobenzoic acid 98%
Pentafluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇HF₅O₂
MDL Number: MFCD00002406
CAS Number: 602-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR