(S)-2-(Cyclohexylamino)propanoic acid 95%

Supplier: Ambeed
Warning

A242318-100MG A242318-1G A242318-250MG A242318-5G
76815-446EA 9 USD
76815-446 76815-448 76815-450 76815-452
(S)-2-(Cyclohexylamino)propanoic acid 95%
(S)-2-(Cyclohexylamino)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MDL Number: MFCD16039417
CAS Number: 82017-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR