tert-Butyl (3R,4S)-3-amino-4-fluoropyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: (3S,4R)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

A242039-100MG A242039-250MG A242039-1G
76815-438EA 103.97 USD
76815-438 76815-440 77568-354
tert-Butyl (3R,4S)-3-amino-4-fluoropyrrolidine-1-carboxylate 95%
tert-Butyl (3R,4S)-3-amino-4-fluoropyrrolidine-1-carboxylate

Precaución: For research use only.

Formula: C₉H₁₇FN₂O₂
MW: 204.25 g/mol
MDL Number: MFCD18071735
CAS Number: 1174020-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR