3-Bromo-2-cyanobenzeneboronic acid 98%

Supplier: Ambeed
Warning

A242034-1G A242034-250MG
76815-434EA 30 USD
76815-434 76815-436
3-Bromo-2-cyanobenzeneboronic acid 98%
3-Bromo-2-cyanobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrNO₂
MDL Number: MFCD10696672
CAS Number: 1032231-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR